1-Ethyl-3-methylimidazolium dicyanamide ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Basionics VS 03, EMIM DCA

E0680-1G E0680-5G
TCE0680-1GEA 36 USD
TCE0680-1G TCE0680-5G
1-Ethyl-3-methylimidazolium dicyanamide ≥97.0% (by HPLC, total nitrogen)
1-Ethyl-3-methylimidazolium dicyanamide

CAS Number: 370865-89-7
Molecular Formula: C8H11N5
Molecular Weight: 177.21
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Clear Liquid
Specific Gravity (20/20): 1.11

Formula: C₈H₁₁N₅
MW: 177.21 g/mol
Density: 1.11 (20 °C)
MDL Number: MFCD08276373
CAS Number: 370865-89-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47814f6a15c0e6a2b80e4f8b904f7942c4d8c849&vid=39f831c025fb6c05fac50fd43c82ec8e26351d53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR