1,2-Epoxycyclopentane ≥97.0%
Supplier: TCI America
Synonyms:
Cyclopentene oxide
E0513-5G
E0513-25G
TCE0513-005GEA
36
USD
TCE0513-005G
TCE0513-25G
1,2-Epoxycyclopentane ≥97.0%
1,2-Epoxycyclopentane
CAS Number: 285-67-6
MDL Number: MFCD00005161
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 10
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: Cyclopentene Oxide, 6-Oxabicyclo[3.1.0]hexane
Formula:
C₅H₈O MW: 84.12 Boiling Pt: 102 °C Density: 0.97 (20 °C) Flash Pt: 10 °C |
MDL Number:
MFCD00005161 CAS Number: 285-67-6 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...