3-Ethoxypropionitrile ≥99.0%

Supplier: TCI America

Synonyms: 3-Ethoxypropanenitrile

E0299-500ML E0299-25ML
TCE0299-500MLEA 78 USD
TCE0299-500ML TCE0299-025ML
3-Ethoxypropionitrile ≥99.0%
3-Ethoxypropionitrile

CAS Number: 2141-62-0
MDL Number: MFCD00001959
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 63
Specific Gravity (20/20): 0.91

Formula: C₅H₉NO
MW: 99.1300 g/mol
Boiling Pt: 171 °C
Density: 0.91 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00001959
CAS Number: 2141-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fb98e1febf079688b7ed012333755d62d0cd535&vid=1fb98e1febf079688b7ed012333755d62d0cd535&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR