2,4-Dichlorophenol ≥98.0% for biochemical research

Supplier: TCI America

Synonyms: 1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene

D3865-1G D3865-5G
TCD3865-1GEA 17 USD
TCD3865-1G TCD3865-5G
2,4-Dichlorophenol ≥98.0% for biochemical research
2,4-Dichlorophenol

[for Biochemical Research]
CAS Number: 120-83-2
MDL Number: MFCD00002169
Molecular Formula: C6H4Cl2O
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 43
Flash Point (°C): 114


Product Categories: Bioscience, Peroxidase Substrates, Analytical Science, Chlorinated Compounds (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: 2,4-DCP

Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 210
Melting Pt: 43 °C
Flash Pt: 114 °C
MDL Number: MFCD00002169
CAS Number: 120-83-2
UN: 2928
ADR: 6.1 II
Merck Index: 13,03098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae26e34f212e2bdb6cadfdf8d6e10607736b9063&vid=b7bfb4a2dba633e891218e808f84f66923c436e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR