O-Dianisidine ≥95.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: 4,4'-Diamino-3,3'-dimethoxybiphenyl, 3,3'-Dimethoxybenzidine

D3864-1G D3864-5G
TCD3864-1GEA 20 USD
TCD3864-1G TCD3864-5G
O-Dianisidine ≥95.0% (by titrimetric analysis) for biochemical research
O-Dianisidine

[for Biochemical Research]
CAS Number: 119-90-4
MDL Number: MFCD00008372
Molecular Formula: C14H16N2O2
Molecular Weight: 244.29
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 138
Flash Point (°C): 206


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 4,4'-Diamino-3,3'-dimethoxybiphenyl, 3,3'-Dimethoxybenzidine, Fast Blue B

Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Melting Pt: 138 °C
Flash Pt: 206 °C
MDL Number: MFCD00008372
CAS Number: 119-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06f8fb67cf7cc962c48a5a8dbbca316052008850&vid=4a65e0285ab104a27fbab9e400bd0a912b3c9622&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR