2,6-Dichloronicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

D3412-5G D3412-25G
TCD3412-5GEA 81 USD
TCD3412-5G TCD3412-25G
2,6-Dichloronicotinic acid ≥98.0% (by GC, titration analysis)
2,6-Dichloronicotinic acid

CAS Number: 38496-18-3
MDL Number: MFCD00075583
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 2,6-Dichloro-3-pyridinecarboxylic Acid

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00075583
CAS Number: 38496-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc1e3b879bb97c44e966ab7ef89c495cda3511fe&vid=dc1e3b879bb97c44e966ab7ef89c495cda3511fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR