Dimethyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
L-Glutamic Acid Dimethyl Ester Hydrochloride, H-Glu(OMe)-OMe.HCl
D3353-5G
D3353-25G
TCD3353-5GEA
39
USD
TCD3353-5G
TCD3353-25G
Dimethyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Dimethyl-L-glutamate hydrochloride
CAS Number: 23150-65-4
Molecular Formula: C7H13NO4
Molecular Weight: 211.64
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 93
Specific rotation [a]20/D: 26 deg (C=5, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Glutamic Acid Dimethyl Ester Hydrochloride, H-Glu(OMe)-OMe.HCl
Formula:
C₇H₁₃NO₄ MW: 211.64 g/mol Melting Pt: 93 °C |
MDL Number:
MFCD00038879 CAS Number: 23150-65-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (N,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...