Diethyl allylphosphonate ≥95.0%

Supplier: TCI America

D3069-5G D3069-1G
TCD3069-5GEA 102 USD
TCD3069-5G TCD3069-1G
Diethyl allylphosphonate ≥95.0%
Diethyl allylphosphonate

CAS Number: 1067-87-4
Molecular Formula: C7H15O3P
Molecular Weight: 178.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 106
Specific Gravity (20/20): 1.03


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Materials Chemistry, Monomers, Monomers
Other Product Names / Synonyms: Allylphosphonic Acid Diethyl Ester

Formula: C₇H₁₅O₃P
MW: 178.1660 g/mol
Boiling Pt: 102 °C
Density: 1.03 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00015134
CAS Number: 1067-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9245a747638b3ad5d4bfc6fe3fbce252625ba6d7&vid=9245a747638b3ad5d4bfc6fe3fbce252625ba6d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR