Borane dimethylamine complex ≥95.0% (by titrimetric analysis)

Supplier: TCI America

D1842-25G D1842-100G
TCD1842-25GEA 76.29 USD
TCD1842-25G TCD1842-100G
Borane dimethylamine complex ≥95.0% (by titrimetric analysis)
Borane dimethylamine complex

CAS Number: 74-94-2
MDL Number: MFCD00051068
Molecular Formula: C2H10BN
Molecular Weight: 58.92
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 37
Flash Point (°C): 43
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Borane - Dimethylamine Complex, DMAB

Formula: C₂H₁₀BN
Melting Pt: 37 °C
Flash Pt: 43 °C
MDL Number: MFCD00051068
CAS Number: 74-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8a71c2570d748ea86e2ed923c13aac0b5b65198&vid=a8a71c2570d748ea86e2ed923c13aac0b5b65198&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR