Chlorodimethyloctylsilane ≥96.0%

Supplier: TCI America

D1827-25ML D1827-100ML
TCD1827-025MLEA 83 USD
TCD1827-025ML TCD1827-100ML
Chlorodimethyloctylsilane ≥96.0%
Chlorodimethyloctylsilane

CAS Number: 18162-84-0
MDL Number: MFCD00000511
Molecular Formula: C10H23ClSi
Molecular Weight: 206.83
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 92
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: n-Octyldimethylchlorosilane

Formula: C₁₀H₂₃ClSi
MW: 206.83 g/mol
Boiling Pt: 220 °C
Density: 0.87 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00000511
CAS Number: 18162-84-0
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae4f658a736087ce26efb30f1ae88eb447d5488a&vid=ae4f658a736087ce26efb30f1ae88eb447d5488a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR