4-(Diethylamino)salicylaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-(Diethylamino)-2-hydroxybenzaldehyde
D1752-250G
D1752-25G
TCD1752-250GEA
130
USD
TCD1752-250G
TCD1752-025G
4-(Diethylamino)salicylaldehyde ≥98.0% (by GC, titration analysis)
4-(Diethylamino)salicylaldehyde
CAS Number: 17754-90-4
MDL Number: MFCD00003326
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 60
Formula:
C₁₁H₁₅NO₂ MW: 193.25 g/mol Melting Pt: 60 °C |
MDL Number:
MFCD00003326 CAS Number: 17754-90-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...