3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,5-Dimethylpiperidine

D1411-500ML D1411-25ML
TCD1411-500MLEA 286.6 USD
TCD1411-500ML TCD1411-25ML
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) ≥98.0% (by GC, titration analysis)
3,5-Dimethylpiperidin (mixture of cis- and trans-isomers)

CAS Number: 35794-11-7
MDL Number: MFCD00005996
Molecular Formula: C7H15N
Molecular Weight: 113.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 32
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Hexahydro-3,5-lutidine, 3,5-Lupetidine

Formula: C₇H₁₅N
MW: 113.20 g/mol
Boiling Pt: 144 °C
Density: 0.83 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00005996
CAS Number: 35794-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=225703443600d270c3d2cd3ee40362a0eb87c405&vid=bbc90737353bd625a5eeff56d59243c9c127fcb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR