1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DPTA-OH

D1330-25G D1330-5G
TCD1330-025GEA 168 USD
TCD1330-025G TCD1330-005G
1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid

CAS Number: 3148-72-9
MDL Number: MFCD00004288
Molecular Formula: C11H18N2O9
Molecular Weight: 322.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: DPTA-OH

Formula: C₁₁H₁₈N₂O₉
MW: 322.27 g/mol
MDL Number: MFCD00004288
CAS Number: 3148-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2bc10de1a5f2535dcc47b27689fe9a30bcb3741&vid=d2bc10de1a5f2535dcc47b27689fe9a30bcb3741&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR