Methylenediphenyl-4,4'-diisocyanate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 101-68-8
MDL Number: MFCD00036131
Molecular Formula: C15H10N2O2
Molecular Weight: 250.26
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 184
Melting point (°C): 41
Flash Point (°C): 196
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(4-isocyanatophenyl)methane, 4,4'-Methylenebis(phenyl Isocyanate), 4,4'-Diphenylmethane Diisocyanate, MDI
Formula:
C₁₅H₁₀N₂O₂ Boiling Pt: 184 °C Melting Pt: 41 °C Flash Pt: 196 °C |
MDL Number:
MFCD00036131 CAS Number: 101-68-8 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...