N,N-Diisopropylethylenediamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Aminoethyldiisopropylamine, N,N-Bis(isopropyl)ethane-1,2-diamine
D0006-500ML
D0006-25ML
TCD0006-500MLEA
517.8
USD
TCD0006-500ML
TCD0006-25ML
N,N-Diisopropylethylenediamine ≥97.0% (by GC, titration analysis)
N,N-Diisopropylethylenediamine
CAS Number: 121-05-1
MDL Number: MFCD00015015
Molecular Formula: C8H20N2
Molecular Weight: 144.26
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 54
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: 2-Diisopropylaminoethylamine
Formula:
C₈H₂₀N₂ MW: 144.26 g/mol Boiling Pt: 170 °C Density: 0.83 (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00015015 CAS Number: 121-05-1 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...