2-Chloro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2-Chloro-4-methoxybenzeneboronic acid

C2386-1G C2386-5G
TCC2386-1GEA 58 USD
TCC2386-1G TCC2386-5G
2-Chloro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-methoxyphenylboronic acid

CAS Number: 219735-99-6
MDL Number: MFCD03411935
Molecular Formula: C7H8BClO3
Molecular Weight: 186.40
Form: Crystal
Color: White
Melting point (°C): 202


Other Product Names / Synonyms: 2-Chloro-4-methoxybenzeneboronic Acid

Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Melting Pt: 202 °C
MDL Number: MFCD03411935
CAS Number: 219735-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4c1308810576c4bfa1abeafd515fd7f4fd5bd50&vid=c4c1308810576c4bfa1abeafd515fd7f4fd5bd50&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR