2-Chloro-N-(2,6-dimethylphenyl)acetamide ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide
C2307-25G
C2307-5G
TCC2307-25GEA
95
USD
TCC2307-25G
TCC2307-5G
2-Chloro-N-(2,6-dimethylphenyl)acetamide ≥98.0% (by GC, total nitrogen)
2-Chloro-N-(2,6-dimethylphenyl)acetamide
CAS Number: 1131-01-7
MDL Number: MFCD00000926
Molecular Formula: C10H12ClNO
Molecular Weight: 197.66
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 148
Other Product Names / Synonyms: N-Chloroacetyl-2,6-dimethylaniline
Formula:
C₁₀H₁₂ClNO MW: 197.66 g/mol Melting Pt: 148 °C |
MDL Number:
MFCD00000926 CAS Number: 1131-01-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...