3-Chloro-6-methylpyridazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2274-25G C2274-5G
TCC2274-25GEA 402.8 USD
TCC2274-25G TCC2274-5G
3-Chloro-6-methylpyridazine ≥98.0% (by GC, titration analysis)
3-Chloro-6-methylpyridazine

CAS Number: 1121-79-5
MDL Number: MFCD00052905
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 60

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00052905
CAS Number: 1121-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5871495b0a483cf7c551031f39f7d323a7dd9b46&vid=5871495b0a483cf7c551031f39f7d323a7dd9b46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR