2-Hydroxy-6-methylnicotinonitrile ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C2210-5G C2210-25G
TCC2210-5GEA 57 USD
TCC2210-5G TCC2210-25G
2-Hydroxy-6-methylnicotinonitrile ≥98.0% (by titrimetric analysis)
2-Hydroxy-6-methylnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

CAS Number: 4241-27-4
MDL Number: MFCD00006013
Molecular Formula: C7H6N2O
Molecular Weight: 134.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 295


Other Product Names / Synonyms: 5-Cyano-6-hydroxy-2-picoline, 3-Cyano-6-methyl-2-pyridinol, 2-Hydroxy-6-methylnicotinonitrile

Formula: C₇H₆N₂O
MW: 134.14 g/mol
Melting Pt: 295 °C
MDL Number: MFCD00006013
CAS Number: 4241-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e0ab75a85c594040915d637fb3846605ba65d09&vid=3e0ab75a85c594040915d637fb3846605ba65d09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR