2-Chloro-4,6-dimethylpyrimidine ≥98.0%

Supplier: TCI America

C2199-25G C2199-5G
TCC2199-25GEA 179 USD
TCC2199-25G TCC2199-5G
2-Chloro-4,6-dimethylpyrimidine ≥98.0%
2-Chloro-4,6-dimethylpyrimidine

CAS Number: 4472-44-0
MDL Number: MFCD00023199
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 212
Melting point (°C): 37
Storage Temperature: 0-10°C

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 212 °C
Melting Pt: 37 °C
MDL Number: MFCD00023199
CAS Number: 4472-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cfacd12633674fd6cbd749dc1369c631af0dac0&vid=3cfacd12633674fd6cbd749dc1369c631af0dac0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR