N(α)-Cbz-D-tryptophan ≥98.0%

Supplier: TCI America

Synonyms: Z-D-Trp-OH, N(α)-Benzyloxycarbonyl-D-tryptophan

C2130-1G C2130-5G
TCC2130-1GEA 45 USD
TCC2130-1G TCC2130-5G
N(α)-Cbz-D-tryptophan ≥98.0%
N(α)-Cbz-D-tryptophan

CAS Number: 2279-15-4
MDL Number: MFCD00037945
Molecular Formula: C19H18N2O4
Molecular Weight: 338.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 123
Specific rotation [a]20/D: 11 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-D-tryptophan, Z-D-Trp-OH

Formula: C₁₉H₁₈N₂O₄
Melting Pt: 123 °C
MDL Number: MFCD00037945
CAS Number: 2279-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c9921f9271f7615d5aa9955afee7fe70b6c171d&vid=0c9921f9271f7615d5aa9955afee7fe70b6c171d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR