3-Cyanophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid
C2031-25G
C2031-5G
TCC2031-25GEA
144
USD
TCC2031-25G
TCC2031-5G
3-Cyanophenylboronic acid (contains varying amounts of Anhydride)
3-Cyanophenylboronic acid
CAS Number: 150255-96-2
Molecular Formula: C7H6BNO2
Molecular Weight: 146.94
Form: Crystal
Color: White
Melting point (°C): 298
Other Product Names / Synonyms: 3-Cyanobenzeneboronic Acid
Formula:
C₇H₆BNO₂ MW: 146.94 g/mol Melting Pt: 298 °C |
MDL Number:
MFCD01318967 CAS Number: 150255-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...