6-Chlorooxindole ≥98.0%
Supplier: TCI America
Synonyms:
6-Chloro-2-oxindole
C2024-25G
C2024-5G
TCC2024-25GEA
91
USD
TCC2024-25G
TCC2024-5G
6-Chlorooxindole ≥98.0%
6-Chlorooxindole
CAS Number: 56341-37-8
Molecular Formula: C8H6ClNO
Molecular Weight: 167.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 197
Other Product Names / Synonyms: 6-Chloro-2-indolinone
Formula:
C₈H₆ClNO MW: 167.59 g/mol Melting Pt: 197 °C |
MDL Number:
MFCD00209962 CAS Number: 56341-37-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...