5-Chloro-2-nitrobenzamide ≥90.0%

Supplier: TCI America

C1978-25G C1978-5G
TCC1978-25GEA 577 USD
TCC1978-25G TCC1978-5G
5-Chloro-2-nitrobenzamide ≥90.0%
5-Chloro-2-nitrobenzamide

CAS Number: 40763-96-0
Molecular Formula: C7H5ClN2O3
Molecular Weight: 200.58
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Melting point (°C): 154

Formula: C₇H₅ClN₂O₃
MW: 200.58 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00017509
CAS Number: 40763-96-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f4f53af4b57b581b34bd88d047f477b906217c5&vid=6f4f53af4b57b581b34bd88d047f477b906217c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR