1-Cyclopentene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-Cyclopentenecarboxylic acid

C1702-1G C1702-5G
TCC1702-1GEA 86 USD
TCC1702-1G TCC1702-5G
1-Cyclopentene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-Cyclopentene-1-carboxylic acid

CAS Number: 1560-11-8
MDL Number: MFCD00001396
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 122

Formula: C₆H₈O₂
MW: 112.13 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00001396
CAS Number: 1560-11-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e5ef7dd99bef0def966112d3fc70231f2a863da&vid=7e5ef7dd99bef0def966112d3fc70231f2a863da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR