Methyl cyclopropanecarboxylate ≥97.0%
Supplier: TCI America
Synonyms:
Methylcyclopropanecarboxylate
C1607-25ML
C1607-250ML
TCC1607-025MLEA
27
USD
TCC1607-025ML
TCC1607-250ML
Methyl cyclopropanecarboxylate ≥97.0%
Methyl cyclopropanecarboxylate
CAS Number: 2868-37-3
MDL Number: MFCD00001280
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 18
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: Cyclopropanecarboxylic Acid Methyl Ester
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 117 °C Density: 1.00 (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00001280 CAS Number: 2868-37-3 UN: 3272 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...