4-Cyanobenzamide ≥97.0% (by total nitrogen basis)

Supplier: TCI America

C1594-5G C1594-1G
TCC1594-5GEA 139.1 USD
TCC1594-5G TCC1594-001G
4-Cyanobenzamide ≥97.0% (by total nitrogen basis)
4-Cyanobenzamide

CAS Number: 3034-34-2 MDL Number: MFCD00017133 Molecular Formula: C8H6N2O Molecular Weight: 146.15 Purity/Analysis Method: 97.0% (N) Form: Crystal Melting point (°C): 226

Formula: C₈H₆N₂O
Melting Pt: 226 °C
MDL Number: MFCD00017133
CAS Number: 3034-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f832374a1e527aaf432e10df3202db16cf64a404&vid=f832374a1e527aaf432e10df3202db16cf64a404&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR