Ethyl-2-cyano-2-methylpropionate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-Cyano-2-methylpropionate, 2-Cyano-2-methylpropionic Acid Ethyl Ester

C1546-5G C1546-25G
TCC1546-5GEA 43 USD
TCC1546-5G TCC1546-025G
Ethyl-2-cyano-2-methylpropionate ≥98.0%
Ethyl-2-cyano-2-methylpropionate

CAS Number: 1572-98-1
MDL Number: MFCD00034667
Molecular Formula: C7H11NO2
Molecular Weight: 141.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 2-Cyano-2-methylpropionic Acid Ethyl Ester

Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 185 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00034667
CAS Number: 1572-98-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f2e27afad298ca84aa5a2e4a3a25bde269ea967&vid=5f2e27afad298ca84aa5a2e4a3a25bde269ea967&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR