Ethyl-4-oxocyclohexanecarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone

C1530-5G C1530-25G
TCC1530-005GEA 27 USD
TCC1530-005G TCC1530-025G
Ethyl-4-oxocyclohexanecarboxylate ≥98.0%
Ethyl-4-oxocyclohexanecarboxylate

CAS Number: 17159-79-4
MDL Number: MFCD00013285
Molecular Formula: C9H14O3
Molecular Weight: 170.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 110
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 4-Oxocyclohexanecarboxylic Acid Ethyl Ester

Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Boiling Pt: 152 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013285
CAS Number: 17159-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8f0ad7a79543401c8d1b766d430a27ecd5ff52&vid=8a8f0ad7a79543401c8d1b766d430a27ecd5ff52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR