Carbonylhydridotris(triphenylphosphine)rhodium(I)

Supplier: TCI America

Synonyms: Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)

C1383-1G C1383-5G
TCC1383-001GEA 397 USD
TCC1383-001G TCC1383-5G
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Carbonylhydridotris(triphenylphosphine)rhodium(I)

CAS Number: 17185-29-4
MDL Number: MFCD00151644
Molecular Formula: C55H46OP3Rh
Molecular Weight: 918.80
Form: Crystal
Color: Yellow
Melting point (°C): 122


Other Product Names / Synonyms: Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride

Formula: C₅₅H₄₆OP₃Rh
MW: 918.80 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00151644
CAS Number: 17185-29-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e9e98b838c8a802e791258cad5a423d0d2200b6&vid=7e9e98b838c8a802e791258cad5a423d0d2200b6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR