(3-Cyanopropyl)dimethylchlorosilane ≥95.0%
Supplier: TCI America
Synonyms:
Chloro(3-cyanopropyl)dimethylsilane
C1207-25ML
C1207-5ML
TCC1207-025MLEA
276.77
USD
TCC1207-025ML
TCC1207-5ML
(3-Cyanopropyl)dimethylchlorosilane ≥95.0%
(3-Cyanopropyl)dimethylchlorosilane
CAS Number: 18156-15-5
MDL Number: MFCD00001969
Molecular Formula: C6H12ClNSi
Molecular Weight: 161.70
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 108
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Chloro(3-cyanopropyl)dimethylsilane
Formula:
C₆H₁₂ClNSi MW: 161.70 g/mol Boiling Pt: 109 °C Density: 0.99 (20 °C) Flash Pt: 108 °C |
MDL Number:
MFCD00001969 CAS Number: 18156-15-5 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...