2-(1-Cyclohexenyl)ethylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(Cyclohex-1-en-1-yl)ethanamine
C1157-500ML
C1157-25ML
TCC1157-500MLEA
285
USD
TCC1157-500ML
TCC1157-25ML
2-(1-Cyclohexenyl)ethylamine ≥98.0% (by GC, titration analysis)
2-(1-Cyclohexenyl)ethylamine
CAS Number: 3399-73-3
MDL Number: MFCD00012058
Molecular Formula: C8H15N
Molecular Weight: 125.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 57
Specific Gravity (20/20): 0.90
Other Product Names / Synonyms: 2-Aminoethyl-1-cyclohexene
Formula:
C₈H₁₅N MW: 125.2114 g/mol Boiling Pt: 84 °C Density: 0.90 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00012058 CAS Number: 3399-73-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...