6-(Chloromethyl)uracil ≥98.0%
Supplier: TCI America
Synonyms:
6-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione
C1057-5G
C1057-25G
TCC1057-005GEA
49
USD
TCC1057-005G
TCC1057-025G
6-(Chloromethyl)uracil ≥98.0%
6-(Chloromethyl)uracil
CAS Number: 18592-13-7
MDL Number: MFCD00010095
Molecular Formula: C5H5ClN2O2
Molecular Weight: 160.56
Purity/Analysis Method:
Formula:
C₅H₅ClN₂O₂ MW: 160.56 g/mol |
MDL Number:
MFCD00010095 CAS Number: 18592-13-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...