Vinyl chloroacetate ≥99.0% stabilized
Supplier: TCI America
Synonyms:
Chloroacetic Acid Vinyl Ester
C0627-500G
C0627-25G
TCC0627-500GEA
363.27
USD
TCC0627-500G
TCC0627-25G
Vinyl chloroacetate ≥99.0% stabilized
Vinyl chloroacetate
(stabilized with MEHQ)
CAS Number: 2549-51-1
MDL Number: MFCD00045275
Molecular Formula: C4H5ClO2
Molecular Weight: 120.53
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 51
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: Chloroacetic Acid Vinyl Ester
Formula:
C₄H₅ClO₂ MW: 120.53 g/mol Boiling Pt: 136 °C Density: 1.19 (20 °C) Flash Pt: 51 °C |
MDL Number:
MFCD00045275 CAS Number: 2549-51-1 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...