N-Carbobenzoxy-L-glutamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-Cbz-L-glutamine, Z-Gln-OH
C0574-5G
C0574-25G
TCC0574-005GEA
25
USD
TCC0574-005G
TCC0574-025G
N-Carbobenzoxy-L-glutamine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-L-glutamine
CAS Number: 2650-64-8
MDL Number: MFCD00008043
Molecular Formula: C13H16N2O5
Molecular Weight: 280.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 138
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-glutamine, Z-Gln-OH
Formula:
C₁₃H₁₆N₂O₅ MW: 280.28 g/mol Melting Pt: 138 °C |
MDL Number:
MFCD00008043 CAS Number: 2650-64-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...