D-(+)-Cellobiose ≥98.0%
Supplier: TCI America
Synonyms:
β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
CAS Number: 528-50-7
MDL Number: MFCD00136034
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Specific rotation [a]20/D: 34 deg (C=8, H2O)
Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Disaccharides
Other Product Names / Synonyms: D-(+)-Cellose
Formula:
C₁₂H₂₂O₁₁ MW: 342.30 g/mol |
MDL Number:
MFCD00136034 CAS Number: 528-50-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...