5-Bromo-2-(trifluoromethyl)pyridine ≥98.0%
Supplier: TCI America
Synonyms:
5-Bromo-2-trifluoromethylpyridine
B3451-1G
B3451-5G
TCB3451-1GEA
38
USD
TCB3451-1G
TCB3451-5G
5-Bromo-2-(trifluoromethyl)pyridine ≥98.0%
5-Bromo-2-(trifluoromethyl)pyridine
CAS Number: 436799-32-5
MDL Number: MFCD06657686
Molecular Formula: C6H3BrF3N
Molecular Weight: 226.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 78
Melting point (°C): 40
Flash Point (°C): 78
Formula:
C₆H₃BrF₃N MW: 226.00 g/mol Boiling Pt: 78 °C Melting Pt: 40 °C Flash Pt: 78 °C |
MDL Number:
MFCD06657686 CAS Number: 436799-32-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...