3-Bromochromone ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromo-4H-chromen-4-one
B3334-25G
B3334-5G
TCB3334-25GEA
341
USD
TCB3334-25G
TCB3334-5G
3-Bromochromone ≥98.0%
3-Bromochromone
CAS Number: 49619-82-1
MDL Number: MFCD00017337
Molecular Formula: C9H5BrO2
Molecular Weight: 225.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 97
Other Product Names / Synonyms: 3-Bromo-4-oxo-4H-1-benzopyran
Formula:
C₉H₅BrO₂ MW: 225.05 g/mol Melting Pt: 97 °C |
MDL Number:
MFCD00017337 CAS Number: 49619-82-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...