O-tert-Butyl-L-tyrosine ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
H-Tyr(tBu)-OH
B3212-1G
B3212-5G
TCB3212-1GEA
31
USD
TCB3212-1G
TCB3212-5G
O-tert-Butyl-L-tyrosine ≥98.0% (by HPLC)
O-tert-Butyl-L-tyrosine
CAS Number: 18822-59-8
MDL Number: MFCD00037202
Molecular Formula: C13H19NO3
Molecular Weight: 237.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 243
Specific rotation [a]20/D: -6 deg (C=1, 1mol/L HCl)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-Tyr(tBu)-OH
Formula:
C₁₃H₁₉NO₃ MW: 237.30 g/mol Melting Pt: 243 °C |
MDL Number:
MFCD00037202 CAS Number: 18822-59-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...