4-Bromo-N,N-diethylaniline ≥97.0%

Supplier: TCI America

B3027-25G B3027-5G
TCB3027-25GEA 214 USD
TCB3027-25G TCB3027-5G
4-Bromo-N,N-diethylaniline ≥97.0%
4-Bromo-N,N-diethylaniline

CAS Number: 2052-06-4
MDL Number: MFCD00013530
Molecular Formula: C10H14BrN
Molecular Weight: 228.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 215
Flash Point (°C): 110
Freezing point (°C): 32
Storage Temperature: 0-10°C

Formula: C₁₀H₁₄BrN
MW: 228.13 g/mol
Boiling Pt: 215 °C
Flash Pt: 110 °C
MDL Number: MFCD00013530
CAS Number: 2052-06-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a954fc38a345f01f86dc6c438c698c0f6d31624e&vid=a954fc38a345f01f86dc6c438c698c0f6d31624e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR