4-Benzamidopiperidine hydrate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-(4-Piperidyl)benzamide
B3025-5G
B3025-25G
TCB3025-5GEA
63.16
USD
TCB3025-5G
TCB3025-25G
4-Benzamidopiperidine hydrate ≥98.0% (by GC, titration analysis)
4-Benzamidopiperidine hydrate
CAS Number: 33953-37-6
MDL Number: MFCD01314226
Molecular Formula: C12H16N2O
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 137
Other Product Names / Synonyms: N-(4-Piperidyl)benzamide
Formula:
C₁₂H₁₆N₂O MW: 204.27 g/mol Melting Pt: 137 °C |
MDL Number:
MFCD01314226 CAS Number: 33953-37-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...