tert-Butyl-3-hydroxyazetidine-1-carboxylate ≥96.0%
Supplier: TCI America
Synonyms:
N-Boc-3-hydroxyazetidine, tert-Butyl 3-hydroxyazetidine-1-carboxylate, 1-BOC-3-Hydroxyazetidine, tert-butyl 3-hydroxyazetidine-1-carboxylate
B2922-1G
B2922-5G
TCB2922-1GEA
37
USD
TCB2922-1G
TCB2922-5G
tert-Butyl-3-hydroxyazetidine-1-carboxylate ≥96.0%
tert-Butyl-3-hydroxyazetidine-1-carboxylate
CAS Number: 141699-55-0
MDL Number: MFCD04115305
Molecular Formula: C8H15NO3
Molecular Weight: 173.21
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 47
Flash Point (°C): 110
Other Product Names / Synonyms: 1-Boc-3-hydroxyazetidine, tert-Butyl 3-Hydroxyazetidine-1-carboxylate, 3-Hydroxyazetidine-1-carboxylic Acid tert-Butyl Ester
Formula:
C₈H₁₅NO₃ MW: 173.21 g/mol Melting Pt: 47 °C Flash Pt: 110 °C |
MDL Number:
MFCD04115305 CAS Number: 141699-55-0 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...