1-Bromo-4-chloro-2-fluorobenzene ≥98.0%

Supplier: TCI America

B2724-5G B2724-25G
TCB2724-5GEA 26 USD
TCB2724-5G 76219-358
1-Bromo-4-chloro-2-fluorobenzene ≥98.0%
1-Bromo-4-chloro-2-fluorobenzene

CAS Number: 1996-29-8
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 92
Specific Gravity (20/20): 1.73

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 92 °C
Density: 1.73 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00079708
CAS Number: 1996-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b8b4210fb425950e23c14cc1a11b671407dc3a9&vid=7b8b4210fb425950e23c14cc1a11b671407dc3a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR