1-Boc-4-hydroxypiperidine ≥97.0%
Supplier: TCI America
Synonyms:
N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate
B2671-5G
B2671-25G
TCB2671-5GEA
17
USD
TCB2671-5G
TCB2671-25G
1-Boc-4-hydroxypiperidine ≥97.0%
1-Boc-4-hydroxypiperidine
CAS Number: 109384-19-2
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 63
Other Product Names / Synonyms: 1-Boc-4-hydroxypiperidine, 1-Boc-4-piperidinol
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol Melting Pt: 63 °C |
MDL Number:
MFCD01075174 CAS Number: 109384-19-2 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...