1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-Benzyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 1-Benzyl Ester, BOC-L-Aspartic acid 1-benzyl ester, N-Boc-L-aspartic Acid 1-Benzyl Ester, Boc-Asp-OBzl, Boc-Asp-Obzl

B2408-25G B2408-5G
TCB2408-25GEA 154.34 USD
TCB2408-25G TCB2408-5G
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

CAS Number: 30925-18-9
Molecular Formula: C16H21NO6
Molecular Weight: 323.35
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 99
Specific rotation [a]20/D: -24 deg (C=1, MeOH)


Other Product Names / Synonyms: 1-Benzyl N-Boc-L-aspartate, N-Boc-L-aspartic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 1-Benzyl Ester

Formula: C₁₆H₂₁NO₆
MW: 323.35 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00065563
CAS Number: 30925-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e19037d0860f221516ac356c7b719589f25d5d8&vid=5e19037d0860f221516ac356c7b719589f25d5d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR