1-Butyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]
B2195-5G
B2195-25G
TCB2195-5GEA
29.48
USD
TCB2195-5G
TCB2195-25G
1-Butyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazoliumtetrafluoroborate
CAS Number: 174501-65-6
MDL Number: MFCD03095449
Molecular Formula: C8H15BF4N2
Molecular Weight: 226.03
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Clear Liquid
Flash Point (°C): 288
Specific Gravity (20/20): 1.21
Formula:
C₈H₁₅BF₄N₂ MW: 226.02 g/mol Density: 1.21 (20 °C) Flash Pt: 288 °C |
MDL Number:
MFCD03095449 CAS Number: 174501-65-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...