1,2-Bis(chlorodimethylsilyl)ethane ≥95.0%

Supplier: TCI America

B1688-5G B1688-25G
TCB1688-005GEA 55.94 USD
TCB1688-005G TCB1688-025G
1,2-Bis(chlorodimethylsilyl)ethane ≥95.0%
1,2-Bis(chlorodimethylsilyl)ethane

[Protecting Reagent for Primary Amines]
CAS Number: 13528-93-3
MDL Number: MFCD00000505
Molecular Formula: C6H16Cl2Si2
Molecular Weight: 215.26
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 198
Flash Point (°C): 65
Freezing point (°C): 36

Formula: C₆H₁₆Cl₂Si₂
MW: 215.26 g/mol
Boiling Pt: 198 °C
Flash Pt: 65 °C
MDL Number: MFCD00000505
CAS Number: 13528-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2378dfdbcfe102cffe63e61707a34489454dbe92&vid=2378dfdbcfe102cffe63e61707a34489454dbe92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR