Benzyltrimethylammonium dichloroiodate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
B1604-25G
B1604-5G
TCB1604-25GEA
54.96
USD
TCB1604-25G
TCB1604-005G
Benzyltrimethylammonium dichloroiodate ≥97.0% (by titrimetric analysis)
Benzyltrimethylammonium dichloroiodate
CAS Number: 114971-52-7
MDL Number: MFCD00075259
Molecular Formula: C10H16Cl2IN
Molecular Weight: 348.05
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 125
Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Trimethylbenzylammonium Dichloroiodate
Formula:
C₁₀H₁₆Cl₂IN MW: 348.06 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00075259 CAS Number: 114971-52-7 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...