Ethyl-3-(benzylamino)propionate ≥95.0%

Supplier: TCI America

Synonyms: N-Benzyl-beta-alanine Ethyl Ester, N-Benzyl-3-aminopropionic Acid Ethyl Ester, Ethyl 3-(benzylamino)propanoate

B1579-500ML B1579-25ML
TCB1579-500MLEA 488.6 USD
TCB1579-500ML TCB1579-025ML
Ethyl-3-(benzylamino)propionate ≥95.0%
Ethyl-3-(benzylamino)propionate

CAS Number: 23583-21-3
MDL Number: MFCD00085541
Molecular Formula: C12H17NO2
Molecular Weight: 207.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C


Other Product Names / Synonyms: N-Benzyl-beta-alanine Ethyl Ester, N-Benzyl-3-aminopropionic Acid Ethyl Ester

Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Boiling Pt: 135 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00085541
CAS Number: 23583-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0b1ce0c9b136239a6d00066f0f0152e1bd8b918&vid=d0b1ce0c9b136239a6d00066f0f0152e1bd8b918&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR