Ethyl-tert-butylacetate ≥99.0%

Supplier: TCI America

Synonyms: tert-Butylacetic Acid Ethyl Ester, Ethyltert-butylacetate, 3,3-Dimethylbutyric Acid Ethyl Ester, Ethyl 3,3-Dimethylbutyrate

B1510-25ML B1510-100ML
TCB1510-025MLEA 70.1 USD
TCB1510-025ML TCB1510-100ML
Ethyl-tert-butylacetate ≥99.0%
Ethyl-tert-butylacetate

CAS Number: 5340-78-3
MDL Number: MFCD00026911
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 34
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: tert-Butylacetic Acid Ethyl Ester, 3,3-Dimethylbutyric Acid Ethyl Ester, Ethyl 3,3-Dimethylbutyrate

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 145 °C
Density: 0.86 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00026911
CAS Number: 5340-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1b6242842708d34f20e4395e9949af793238d5d&vid=a1b6242842708d34f20e4395e9949af793238d5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR